Phenazopyridine hydrochloride, 99%, analytical standard
Phenazopyridine hydrochloride, 99%, analytical standard
Phenazopyridine hydrochloride, 99%, analytical standard
Thermo Scientific Chemicals

Phenazopyridine hydrochloride, 99%, analytical standard

CAS: 136-40-3 | C11H11N5·ClH | 249.7 g/mol
製品番号(カタログ番号)数量
441921000100 mg
製品番号(カタログ番号) 441921000
価格(JPY)
-
数量:
100 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS136-40-3
IUPAC Namehydrogen 3-(2-phenyldiazen-1-yl)pyridine-2,6-diamine chloride
Molecular FormulaC11H12ClN5
InChI KeyQQBPIHBUCMDKFG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(N=NC2=CC=CC=C2)C(N)=N1
さらに表示
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)Red
Assay>=98.5 to =<101.5 %
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only