1-Aminohydantoin hydrochloride, 90%
1-Aminohydantoin hydrochloride, 90%
1-Aminohydantoin hydrochloride, 90%
Thermo Scientific Chemicals

1-Aminohydantoin hydrochloride, 90%

CAS: 2827-56-7 | C3H5N3O2·ClH | 151.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 442050250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2827-56-7
IUPAC Name1-aminoimidazolidine-2,4-dione hydrochloride
Molecular FormulaC3H6ClN3O2
InChI KeyWEOHANUVLKERQI-UHFFFAOYSA-N
SMILESCl.NN1CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=88 %
Titration Argentometric>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only