4,6-Dimethyl-2-nitroaniline, 97%
4,6-Dimethyl-2-nitroaniline, 97%
4,6-Dimethyl-2-nitroaniline, 97%
Thermo Scientific Chemicals

4,6-Dimethyl-2-nitroaniline, 97%

CAS: 1635-84-3 | C8H10N2O2 | 166.18 g/mol
Have Questions?
Catalog NumberQuantity
4424400505 g
Catalog number 442440050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1635-84-3
IUPAC Name2,4-dimethyl-6-nitroaniline
Molecular FormulaC8H10N2O2
InChI KeyVSRYYONYIUUFFY-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(N)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to orange
Appearance (Form)Solid
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only