4-Amino-2-(trifluoromethyl)benzonitrile, 97%
4-Amino-2-(trifluoromethyl)benzonitrile, 97%
4-Amino-2-(trifluoromethyl)benzonitrile, 97%
Thermo Scientific Chemicals

4-Amino-2-(trifluoromethyl)benzonitrile, 97%

CAS: 654-70-6 | C8H5F3N2 | 186.14 g/mol
製品番号(カタログ番号) 442470010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS654-70-6
IUPAC Name4-amino-2-(trifluoromethyl)benzonitrile
Molecular FormulaC8H5F3N2
InChI KeyPMDYLCUKSLBUHO-UHFFFAOYSA-N
SMILESNC1=CC=C(C#N)C(=C1)C(F)(F)F
さらに表示
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point138°C to 146°C
HPLC>=96.0 %
Appearance (Color)White to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only