DS-36 Matrix Standard (Dye Set J6) - FAQs
DS-36 Matrix Standard (Dye Set J6) - FAQs
View additional product information for DS-36 Matrix Standard (Dye Set J6) - FAQs (4425042)
1 product FAQs found
What is the TrueMark MSI Assay?
The TrueMark MSI Assay (Cat. No. A45295) is a low-input, single-amplification solution for the rapid identification of microsatellite instability (MSI) in formalin-fixed paraffin-embedded (FFPE) samples from multiple tumor tissue types for research applications. It uses a 6-dye (J6) dye set, the DS-36 Matrix Standard (Dye Set J6) (Cat. No. 4425042) and is compatible with the SeqStudio and 3500 Series genetic analyzers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.