Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Product Image
Applied Biosystems™

TaqMan™ Array Rat Endogenous Control Plate, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, orRead more
Have Questions?
Catalog NumberQuantity
44266971 x 96 well plate
Catalog number 4426697
Price (USD)
273.00
Each
Quantity:
1 x 96 well plate
Price (USD)
273.00
Each
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The first aspect of any experiment looking at relative quantitation of gene expression should be the selection of endogenous control gene(s), to normalize for variations in sample input. The Applied Biosystems® TaqMan® Array Rat Endogenous Control 96-well Plate contains 32 TaqMan® Gene Expression Assays, plated in triplicate, that target genes that have been shown to be good candidates for such an experiment. The plate contains assays to 16 commonly used housekeeping genes. The panel also contains assays to 16 rat orthologs of human genes that have been shown to be expressed constitutively and at moderate abundance across most test samples.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format96-well plate
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 10 μL Reactions
Product LineTaqMan
Quantity1 x 96 well plate
SpeciesRat
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedFast
Unit SizeEach
Contents & Storage
Control Primer-Probe Set(s)

Frequently asked questions (FAQs)

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.

Can poly A+ purified RNA samples be used with the TaqMan Human Endogenous Control Plate (Cta. No. 4309199)?

No. The plate has been designed to evaluate only total RNA. Ribosomal RNA will be removed from poly A+ purified RNA samples, and is required for accurate detection of the 18s rRNA endogenous control assay used on the plate.