N-BOC-1,3-diaminopropane, 97%
N-BOC-1,3-diaminopropane, 97%
N-BOC-1,3-diaminopropane, 97%
Thermo Scientific Chemicals

N-BOC-1,3-diaminopropane, 97%

CAS: 75178-96-0 | C8H19N2O2 | 175.25 g/mol
Quantity:
1 mL
5 mL
Packaging:
Glass Bottle
Catalog number 442730010
Price (JPY)
-
Quantity:
1 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS75178-96-0
IUPAC Nametert-butyl N-(3-azaniumylpropyl)carbamate
Molecular FormulaC8H19N2O2
InChI KeyPOHWAQLZBIMPRN-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)NCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only