L-Alanine tert-butyl ester hydrochloride, 99%
L-Alanine tert-butyl ester hydrochloride, 99%
L-Alanine tert-butyl ester hydrochloride, 99%
Thermo Scientific Chemicals

L-Alanine tert-butyl ester hydrochloride, 99%

CAS: 13404-22-3 | C7H16ClNO2 | 181.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 442920050
Price (USD)
140.65
Special offer
Online exclusive
Ends: 28-Dec-2026
165.00
Save 24.35
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
140.65
Special offer
Online exclusive
Ends: 28-Dec-2026
165.00
Save 24.35
Each
Chemical Identifiers
CAS13404-22-3
IUPAC Nametert-butyl (2S)-2-aminopropanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyWIQIWPPQGWGVHD-JEDNCBNOSA-N
SMILESCl.C[C@H](N)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
Titration Argentometric>=98.5 to =<101.5 %
Appearance (Color)White to off-white
Specific optical rotation+1.2° to +1.6° (20°C, 589 nm) (c=2 in EtOH)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only