2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard
Thermo Scientific Chemicals

2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard

CAS: 19767-45-4 | C2H5NaO3S2 | 164.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number 443150050
Price (EUR)
46,65
Online exclusive
52,10
Save 5,45 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
46,65
Online exclusive
52,10
Save 5,45 (10%)
Each
Specifications
Assay Percent Range97.5 to 102% min. (On dried substance) (Iodometry)
CAS19767-45-4
Chemical Name or Material2-Mercaptoethanesulfonic acid sodium salt
EINECS Number243-285-9
Formula Weight164.18
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only