Product Image
Applied Biosystems™

Region of Interest (ROI) and Background Plates, 384-well

These Region-of-Interest (ROI) and Background Plates are used to maintain the ViiA™ 7, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7, andRead more
Have Questions?
Catalog NumberQuantity
44323202 plates
Catalog number 4432320
Price (MXN)
-
Quantity:
2 plates
These Region-of-Interest (ROI) and Background Plates are used to maintain the ViiA™ 7, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7, and QuantStudio™ 12K Flex real-time PCR systems with 384-well blocks. See the instrument maintenance guide for full instructions on use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeFAM, LIZ, NED, ROX, VIC
Calibration TypeRegion-of-Interest (ROI) Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA™ 7 System
Format384-well Plate
Product LineViiA
Quantity2 plates
Shipping ConditionDry Ice
Product TypeBackground Plate
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C

Frequently asked questions (FAQs)

I cannot open the QuantStudio 3/5 Design & Analysis 2 (DA2) software on my Mac. What are the software requirements for Mac?

To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:

Safely open apps on your mac

Please note, the software should run on Mac OS 10.10 or later.

Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center.

How can I get a summary of the calibrations performed on the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Do I need to back up my calibration files before I upgrade the software for the ViiA7 Real-Time PCR System?

No, you do not need to back up the calibration files as all of the calibration files are stored on the ViiA7 Real-Time PCR instrument. Once the computer software is properly installed and the computer is connected to the instrument, the calibration information will automatically update in the QuantStudio Software.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can I still start a run on the ViiA7 Real-Time PCR System if the "Calibration is expired" notification is displayed?

Yes, you can still start a run on the ViiA7 Real-Time PCR System if the expired calibration notice displays. This is just a reminder and you can close it to proceed with the run. Additionally, you can change when you want the notifications to show up and how frequently you want to be notified to calibrate the instrument. To do this, go to the toolbar in the QuantStudio Software and select Instrument > Instrument Console. Select your instrument icon and click on Manager Instrument > Maintenance > Calibration Reminders. From the Calibration reminders Settings page you can update when the calibrations expire, turn expiration reminders on/off, and set when the reminders show up.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.