ROX™ Dye Calibration Plate, 384-well
ROX™ Dye Calibration Plate, 384-well
Applied Biosystems™

ROX™ Dye Calibration Plate, 384-well

The 384-Well Dye Calibration Plate (ROX™) is used to maintain your real-time PCR instrument with 384-well block. Dye calibration shouldRead more
Have Questions?
Catalog NumberQuantity
44324581 x 384 well plate
Catalog number 4432458
Price (MXN)
-
Quantity:
1 x 384 well plate
The 384-Well Dye Calibration Plate (ROX™) is used to maintain your real-time PCR instrument with 384-well block. Dye calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.
Specifications
Calibrated DyeROX, ROX™
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA™ 7 Dx System, QuantStudio™ Dx
Format384-well Dye Calibration Plate
PCR MethodqPCR
Quantity1 x 384 well plate
Shipping ConditionDry Ice
Product TypeDye Calibration Plate
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.