Product Image
Applied Biosystems™

ROI & Background Calibration Kit, Fast 96-well

The Fast 96-Well ROI & Background Calibration Kit is used to maintain your real-time PCR instrument with Fast 96-well block.Read more
Catalog NumberQuantity
44326382 x 96 well plates
Catalog number 4432638
Price (JPY)
45,900
Each
Quantity:
2 x 96 well plates
The Fast 96-Well ROI & Background Calibration Kit is used to maintain your real-time PCR instrument with Fast 96-well block. Region of interest (ROI) and background calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.
Specifications
Calibrated DyeProprietary Mix
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA™ 7 Dx System, QuantStudio™ Dx
PCR MethodqPCR
Quantity2 x 96 well plates
Reaction SpeedFast
Shipping ConditionDry Ice
Product TypeBackground Calibration Kit
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.