Clindamycin hydrochloride, 97%
Clindamycin hydrochloride, 97%
Clindamycin hydrochloride, 97%
Thermo Scientific Chemicals

Clindamycin hydrochloride, 97%

Clindamycin hydrochloride, CAS # 21462-39-5, is a lincosamide antibiotic. | CAS: 21462-39-5 | C18H34Cl2N2O5S | 461.439 g/mol
Have Questions?
Catalog NumberQuantity
44351010010 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 443510100
Price (USD)
76.65
Online exclusive
85.30
Save 8.65
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
76.65
Online exclusive
85.30
Save 8.65
Each
Chemical Identifiers
CAS21462-39-5
IUPAC Namehydrogen (2S,4R)-N-{2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl}-1-methyl-4-propylpyrrolidine-2-carboxamide chloride
Molecular FormulaC18H34Cl2N2O5S
InChI KeyAUODDLQVRAJAJM-GQYOKNPUSA-N
SMILES[H+].[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)NC(C(C)Cl)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water=<6 %
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Clindamycin is a lincosamide antibiotic derived from lincomycin that has antibiotic activity against aerobic gram-positive cocci, and parasitic protozoa
  • Clindamycin reversibly binds to the 50s bacterial ribosomal subunit, thus inhibiting protein synthesis

Applications

  • Clindamycin has been used to enhance bacterial colonization in research mice during the study of commensal gut microbiota
  • Clindamycin has been used to study biofilm-mediated antibiotic resistance in an in vitro model of G. vaginalis
  • Clindamycin was used as a selection agent to isolate, enumerate, and identify Lactobacillus spp. from probiotic dairy products
RUO – Research Use Only

General References:

  1. Spížek, J.; Řezanka, T. Lincosamides: Chemical structure, biosynthesis, mechanism of action, resistance, and applications. Biochem Pharmacol. 2017, 133:20-28.
  2. Patterson, L.; Allen, J.; Posey, I.; Shaw, J.J.P.; Costa-Pinheiro P, Walker SJ, Gademsey A, Wu X, Wu S, Zachos NC, Fox TE, Sears CL, Kester M. Glucosylceramide production maintains colon integrity in response to Bacteroides fragilis toxin-induced colon epithelial cell signaling. FASEB J. 2020, 34(12), 15922-15945.
  3. Farahmand, N.; Ouoba, L.I.I.; Naghizadeh Raeisi, S.; Sutherland, J.; Ghoddusi, H.B. Probiotic Lactobacilli in Fermented Dairy Products: Selective Detection, Enumeration and Identification Scheme. Microorganisms. 2021, 9(8), 1600.