Sequencing Analysis Software v5.3.1 - FAQs
Sequencing Analysis Software v5.3.1 - FAQs
View additional product information for Sequencing Analysis Software v5.3.1 - FAQs (4440845)
8 product FAQs found
We are upgrading to the Windows 10 operating system. What are the compatible capillary electrophoresis secondary analysis software with Windows 10?
The following capillary electrophoresis secondary analysis software are compatible with Windows 10 (and also Windows 7):
-Sequencing Analysis Software v7
-SeqScape Software v4
-Variant Reporter Software v3
-GeneMapper Software v6
-Minor Variant Finder v1.2
-MicrobeBridge v1.1
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where do I get the latest Applied Biosystems Capillary Electrophoresis Software patches?
Please visit our Applied Biosystems Genetic Analyzer Freeware and Patches website: https://www.thermofisher.com/sangerpatches
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We just upgraded to Sequencing Analysis Software v.5.3.1 and I'm wondering if my old data files will work on it?
The Sequencing Analysis Software v.5.3.1 is compatible with the following data files:
310 PC, Data Collection Software v.3.0
3100 series, Data CollectionSoftware v,1.0, 1.1, 2.0
3130 series, Data CollectionSoftware v.3.0
3730 series, Data Collection Software v.3.0
For more information, please click here for the Sequencing Analysis Software v.5.3.1 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Sequencing-Analysis-Software-v5-3-1-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I have a copy of Sequencing Analysis Software v.5.3.1 and want to know what the computer requirements are.
The computer requirements are as follows:
-Minimum: 733 MHz Pentium IV CPU or faster
-512 MB RAM
-5 GB hard disk space for application
For the operating system, Sequencing Analysis Software v.5.3.1 requires Windows 2000, service pack 3 or 4, Windows XP Professional, service pack 2 or 3, or Windows Vista (32-bit Business Edition). Please click here for the Sequencing Analysis Software v.5.3.1 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Sequencing-Analysis-Software-v5-3-1-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We are upgrading to the Windows 7 operating system. Can I install Sequencing Analysis Software v.5.3.1 on that?
Unfortunately, Sequencing Analysis Software v.5.3.1 is not Windows 7 compatible. Please click here for the Sequencing Analysis Software v.5.3.1 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Sequencing-Analysis-Software-v5-3-1-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We just upgraded to Sequencing Analysis Software v.5.4 and I'm wondering if my old data files will work on it?
Sequencing Analysis Software v.5.4 is compatible with the following data files:
310 PC, Data Collection Software v.3.0, 3.1
3100 series, Data Collection Software v.1.0, 1.1, 2.0
3130 series, Data Collection Software v.3.0, 3.1, 4
3730 series, Data Collection Software v.3.0, 3.1, 4
3500 series, Data Collection Software v.1.0, v1.1, 2
For more information, please click here for the Sequencing Analysis Software v.5.4 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Sequencing-Analysis-Software-v-5-4-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I have a copy of Sequencing Analysis Software v.5.4 and want to know what the computer requirements are.
The computer requirements are as follows:
-Minimum: 2.8 GHz Pentium IV CPU or faster
-1 GB RAM
-40 GB hard disk space for application
Recommended CPU: Cor -2 GB RAM -320 GB hard disk space
For the operating system, Sequencing Analysis Software v.5.4 requires either Windows XP Professional, service pack 2 or 3, or Windows Vista (32-bit Business Edition), service pack 1. Please click here for the Sequencing Analysis Software v.5.4 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Sequencing-Analysis-Software-v-5-4-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We are upgrading to the Windows 7 operating system. Can I install Sequencing Analysis Software v.5.4 on that?
Unfortunately, Sequencing Analysis Software v.5.4 is not Windows 7 compatible. Please click here for the Sequencing Analysis Software v.5.4 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Sequencing-Analysis-Software-v-5-4-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.