Ethanol, 100%, technical, denaturated with 5% methanol
Thermo Scientific Chemicals

Ethanol, 100%, technical, denaturated with 5% methanol

CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
產品號碼QuantityPackaging
4441600505 LPlastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 444160050
價格 (TWD)
-
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
化學識別
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
檢視更多
規格Specification Sheet規格表
Refractive index1.3590 to 1.3610 (20°C, 589 nm)
Density(20°C) 0.788 to 0.790 kg/l
GC=<5 % (methanol)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use