Paraffin, liquid, technical
Paraffin, liquid, technical
Paraffin, liquid, technical
Thermo Scientific Chemicals

Paraffin, liquid, technical

CAS: 8012-95-1 | MFCD00131611
Have Questions?
Catalog NumberQuantityPackaging
44418025025 LPlastic Can
Catalog number 444180250
Price (USD)
-
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Chemical Identifiers
CAS8012-95-1
Molecular FormulaMFCD00131611
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4600 to 1.4800 (20°C, 589 nm)
Viscosity20 to 80 mm²/s (40°C)
Infrared spectrumConforms
Appearance (Form)Oily liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only