Applied Biosystems™

SOLiD™ Fragment Library Construction Reagents

Catalog number:  4443713

 Related applications:

SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SOLiD™ Fragment Library Construction Reagents provide a fully optimized set of reagents to generate 10 fragment libraries using the SOLiD™ version 3 standard or express fragment library protocols. This kit replaces S3100105. Additional columns and gels recommended in the SOLiD™ version 3 and 3 plus protocol are available in the SOLiD™ Fragment Library Construction Kits. The SOLiD™ Fragment Library Oligo Kit or an equivalent alternative is also required to generate fragment libraries.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): SOLiD™
Library Type: Fragment Library
Workflow Step: Library Generation
Starting Material: DNA
Sequencing Type: Epigenetic Sequencing, Genome & DNA Sequencing
Downstream Application: Next-Generation Sequencing
Product Line: SOLiD™
Product Size: 10 preps
Shipping Condition: Dry Ice

Contents & storage

SOLiD™ Fragment Library Construction Reagents store at -20 C includes; 5x End Polishing Buffer, dNTP, End Polishing Enzyme 1, End Polishing Enzyme 2, 5x Ligase Buffer, T4 DNA Ligase, Platinum® PCR Amplification Mix

Documents

Manuals & protocols