4-Acetylphenylboronic acid pinacol ester, 97%
4-Acetylphenylboronic acid pinacol ester, 97%
4-Acetylphenylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

4-Acetylphenylboronic acid pinacol ester, 97%

CAS: 171364-81-1 | C14H19BO3 | 246.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444430050
Price (USD)
338.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
338.00
Each
Chemical Identifiers
CAS171364-81-1
IUPAC Name1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-one
Molecular FormulaC14H19BO3
InChI KeyBATKIZWNRQGSKE-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
GC>=96.0 %
NMR (1H-NMR)Consistent with structure
Infrared spectrumConforms
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only