4-Amino-2-fluorophenylboronic acid pinacol ester, 97%
4-Amino-2-fluorophenylboronic acid pinacol ester, 97%
4-Amino-2-fluorophenylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

4-Amino-2-fluorophenylboronic acid pinacol ester, 97%

CAS: 819057-45-9 | C12H17BFNO2 | 237.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444490050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS819057-45-9
IUPAC Name3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H17BFNO2
InChI KeyFLMNWVXAEGUVNY-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C(F)C=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
GC>=96.0 %
Melting point161°C to 167°C
NMR (1H-NMR)Consistent with structure
Appearance (Form)Solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only