2-Chloropyrimidine-5-boronic acid pinacol ester, 95%
2-Chloropyrimidine-5-boronic acid pinacol ester, 95%
2-Chloropyrimidine-5-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

2-Chloropyrimidine-5-boronic acid pinacol ester, 95%

CAS: 1003845-08-6 | C10H14BClN2O2 | 240.49 g/mol
Have Questions?
Catalog NumberQuantity
4447000101 g
Catalog number 444700010
Price (USD)
145.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
145.00
Each
Chemical Identifiers
CAS1003845-08-6
IUPAC Name2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Molecular FormulaC10H14BClN2O2
InChI KeyVLAPDEKXZLRRKV-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
Appearance (Form)Solid or crystalline solid
Infrared spectrumConforms
Melting point63°C to 69°C
GC>=94.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only