4-Iodo-2-methoxyaniline, 97%
4-Iodo-2-methoxyaniline, 97%
4-Iodo-2-methoxyaniline, 97%
Acros Organics

4-Iodo-2-methoxyaniline, 97%

CAS: 338454-80-1 | C7H8INO | 249.05 g/mol
Have Questions?
Catalog NumberQuantity
444800050
also known as 44480
5 g
Catalog number 444800050
also known as 44480
HSN 29222990
Price (INR)
166,016.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
166,016.00
Each
Chemical Identifiers
CAS338454-80-1
IUPAC Name4-iodo-2-methoxyaniline
Molecular FormulaC7H8INO
InChI KeyAEPCMLLYVXZOLQ-UHFFFAOYSA-N
SMILESCOC1=C(N)C=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)Consistent with structure
Appearance (Color)Brown
Appearance (Form)Solid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only