TaqMan™ Gene Expression Assay, VIC
Applied Biosystems™

TaqMan™ Gene Expression Assay, VIC

Applied Biosystems TaqMan Gene Expression assays, VIC, are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5' end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3' end.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4448489S (360 reactions), made to order
4448490M (750 reactions), made to order
4448491L (2900 reactions), made to order
Catalog number 4448489
Price (USD)
408.65
Online exclusive
435.00
Save 26.35
Each
Quantity:
S (360 reactions), made to order

Applied Biosystems TaqMan Gene Expression assays, VIC, are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5' end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3' end.

Features include:

  • Easy to use—just add cDNA and master mix and run—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Extensive content—over 1.8 million predesigned assays available for over 25 different species
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Ideal for multiplexing—combine one FAM-labeled and one VIC-labeled assay to create a duplex assay for two different gene targets in the same reaction

Approximate ship time

Made to order: 4–6 days in North America, 6–10 days in Europe

Pre-formulated assay

  • 2 unlabeled PCR primers (900 nM each final 1X concentration)
  • 1 VIC dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents. All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays Guarantee.*

*Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)60 X
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
FormatTube
No. of Reactions360 reactions
Product LineTaqMan
Product TypeGene Expression Assay
QuantityS (360 reactions), made to order
Shipping ConditionRoom Temperature
Concentration20X
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

Can I multiplex my assays if my amplicons are not all the same length?

Yes, it is possible. The amplicons do not have to be the same length in order to duplex the assays. You need to use different reporter dyes on the probes in multiplex - they should have a substantial difference in maximum emission wavelength to be effective. A classic example is to use FAM and VIC for the 2 probes. Depending on the expression of the targets, you may also need to limit the primer concentration for one of the assays. This is common when duplexing a target and endogenous control (the control probe is VIC labeled and the primer concentration is decreased so the target assay can properly compete for use of enzyme and dNTPs).

How do I determine if Applied Biosystems TaqMan Assays that I want to multiplex are compatible with each other?

You can utilize the Applied Biosystems TaqMan Multiplex Interaction Check (MIC) tool. The MIC tool uses bioinformatics to identify potential interactions between primers, probes, and amplicons that could negatively impact multiplex qPCR performance. The tool was developed and validated using data from approximately 200 multiplex panels, combining computational analysis with wet-lab testing. By flagging assay combinations that may be prone to false positives or reduced sensitivity, MIC tool helps researchers design multiplex panels with greater confidence and reduce time spent on trial-and-error optimization.risk and observed multiplex performance.

Why are the concentrations of forward and reverse primer in our Primer-Limited (PL) Applied Biosystems TaqMan Gene Expression Assays?

Primer-Limited Applied Biosystems TaqMan Gene Expression Assays with FAM, ABY, JUN, or Cyanine 5 dyes have a primer concentration of 350 nM. Assays with VIC dyes have a primer concentration of 150 nM.

Why are some Applied Biosystems TaqMan Gene Expression Assays only available in Primer-Limited concentrations?

In a multiplex qPCR reaction, like in Applied Biosystems TaqMan Gene Expression Assays, targets often differ in abundance and amplification efficiency. Highly expressed or highly efficient targets can consume a disproportionate share of reaction components, generate very strong fluorescent signals, and outcompete weaker targets for polymerase and other reagents. Primer-limited assays provide balanced amplification of targets in multiplex workflows by reducing primer competition between assays and supporting more consistent amplification across multiple targets.

How many Applied Biosystems TaqMan Gene Expression Assays can I multiplex in a single reaction?

You can multiplex up to five Applied Biosystems TaqMan Gene Expression Assays utilizing FAM, VIC, ABY, JUN, and Cyanine 5 dyes.