GeneMapper® ID-X Software v1.2 Full Upgrade
Catalog number: 4448530
Applied Biosystems™  Related applications: Forensics
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This kit includes the GeneMapper® ID-X v1.2 Full Upgrade CD for GeneMapper® ID-X v1.0, 1.0.1, v1.1. and v1.1.1 Full installation upgrade. Documentation is not included.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): 310 Genetic Analyzer, 3100-Avant™ Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
Operating System: Windows 2000, Windows Vista, Windows XP
Product Line: Applied Biosystems™, GeneMapper®
Product Size: 1 license
Software Category: Upgrade
Software Type: Forensic STR Fragment Analysis
Documents
Manuals & protocols