4-Aminocarbonylphenylboronic acid, 97%
4-Aminocarbonylphenylboronic acid, 97%
4-Aminocarbonylphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Aminocarbonylphenylboronic acid, 97%

CAS: 123088-59-5 | C7H8BNO3 | 164.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 444900250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS123088-59-5
IUPAC Name(4-carbamoylphenyl)boronic acid
Molecular FormulaC7H8BNO3
InChI KeyGNRHNKBJNUVWFZ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to tan
Appearance (Form)Solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only