MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit, compatible with PAXgene™ Blood RNA Tubes
Invitrogen™

MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit, compatible with PAXgene™ Blood RNA Tubes

The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit purifies total RNA (including miRNA) from PAXgene™ Blood RNA Tubes using자세히 알아보기
Have Questions?
카탈로그 번호수량
445189496 preps
카탈로그 번호 4451894
제품 가격(KRW)
1,349,000
온라인 행사
Ends: 31-Dec-2026
1,586,000
할인액 237,000 (15%)
Each
수량:
96 preps
제품 가격(KRW)
1,349,000
온라인 행사
Ends: 31-Dec-2026
1,586,000
할인액 237,000 (15%)
Each
The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit purifies total RNA (including miRNA) from PAXgene™ Blood RNA Tubes using proven magnetic bead-based technology. A TURBO DNase™ step efficiently eliminates contaminating gDNA ensuring the highest purity RNA possible suitable for real-time RT-PCR and microarray analysis.

• Flexible purification format—use microfuge tubes, manual 96-well plates, or open-format automated 96-well plates from the same kit
• Simple magnetic bead workflow—no spin columns
• Total RNA without RNase activity, contaminating gDNA, or RT-PCR inhibitors

Maximize Total RNA Yields From Blood Samples
The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit utilizes proven magnetic bead-based technology to purify maximal yields of total RNA from PAXgene™ Blood RNA Tubes. The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit can recover 3-18 μg from a single PAXgene™ Blood RNA tube. A TURBO DNase™ step efficiently eliminates contaminating gDNA that can interfere with downstream applications and ensures the highest purity RNA possible.

The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit performs exceptionally well at purifying miRNA with total RNA from PAXgene™ Blood RNA Tubes without having to employ a separate miRNA protocol. One protocol does it all.

The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit can also detect miRNA from PAXgene™ Blood RNA tubes as shown by TaqMan™ gene expression assays. Users are strongly advised to perform appropriate validation studies against their miRNA targets.

Flexible Purification Formats
The MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit can be processed in low-, medium- or high-throughput formats with superior workflow and handling compared to traditional competitive methods.
• For low-throughput users, the MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit can be used in individual 1.5mL microfuge tubes with the bead RNA-complex remaining in a single processing tube until the final elution step.
• For medium-to-high–throughput users, the MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit can be done in a 96-well processing plate with either a single or multi-channel pipettor to accelerate the purification process.
• For high-throughput users, the MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit is compatible with manual and automated 96-well processing. Simply process multiple batches of tubes in series or in parallel, through the up-front centrifugation process, to the convenient stopping point, until enough samples can be loaded onto a 96-well processing plate for the RNA purification steps. The RNA purification process can be handled on either open-format liquid handlers, in conjunction with a 96-well magnetic stand on the deck, or magnetic bead movers like the MagMAX™ Express-96 Magnetic Particle Processor (deep well version only).

Rapid Processing Time
Processing times will vary based on format selected, but twelve PAXgene™ Blood RNA tubes can be processed manually with the MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit in approximately 2 hours.

An Alternative to Full Purification
For users who perform primarily TaqMan™-based real-time RT-PCR experiments, and do not need to archive RNA⁄cDNA, an alternative to the MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit is the Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR. This kit makes it possible to perform expression analysis directly from PAXgene™ Blood RNA tubes without conventional RNA purification.

One-Stop Shopping
MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit is compatible with our suite of reverse transcription mixes (SuperScript™), PCR master mixes (both TaqMan™ and SYBR™ based), and other blood workflow products. Simplify the ordering process and reduce freight cost by getting more from one supplier.

Specifications:
• Yield (Range): 3 - 18μg
• Quality (RIN) ): ≥ 8
• Quality (A260⁄280) ): 1.8 – 2.2
• gDNA (w⁄w) ): less than 1.56%
• Equipment Needed :
   *Refrigerated centrifuge (4°C) with swing bucket rotor capable of 5,000 x g with
   *50 mL conical tube adapters
    *50ml conical tubes
   *For tube format users: Tabletop microcentrifuge or picocentrifuge
• Number of Preps: 96 preparations (blood tubes)

NOTE: Blood tubes and magnetic stands are sold separately.

NOTE: Blood tubes and magnetic stands are sold separately.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

PAXgene™ and PreAnalytiX™ are registered trademarks of PreAnalytiX GmbH. No sponsorship, endorsement, or affiliation is implied herein.
For Research Use Only. Not for use in diagnostic procedures.
사양
용리량20 to 80 μL
최종 제품 유형Total RNA
용도(애플리케이션)RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
고처리량 호환성High-throughput Compatible
정제 시간Less than 2 hr. when using automated protocol
수량96 preps
배송 조건Box 1: Room Temperature
Box 2: Cold Gel
시작 물질 양2.5 mL of blood in PAXgene™ tube
수율≤18 μg
Isolation TechnologyMagnetic Bead
샘플 종류Whole Blood
Unit SizeEach
구성 및 보관

Box 1
• 2 mL RNA Binding Beads; 4°C
• 18 mL Resuspension Solution; room temperature
• 16 mL Wash Solution 1 (concentrated); room temperature
• 2 x 30 mL Wash Solution 2 (concentrated); room temperature
• 10 mL RNA Rebinding Buffer; room temperature
• 10 mL Elution Buffer; room temperature
• 1 Processing Plate; room temperature
• 100 Tubes; room temperature

Box 2
• 0.6 mL Proteinase; -20°C
• 1 mL TURBO DNase; -20°C
• 1 mL 10X TURBO DNase Buffer; -20°C

자주 묻는 질문(FAQ)

Are MagMax kits compatible with RNAlater reagent-treated samples?

Only MagMax96 for Microarrays Total RNA Isolation Kit (Cat. No. AM1839) is compatible, since it uses Trizol reagent and most of the salt carry-over from RNAlater reagent is eliminated during the lysis steps and partitioned from the aqueous RNA layer on top. For other MagMax kits, optimization is needed. High salt carry over is not optimal for MagMax RNA isolation steps.

Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.

I am getting MagMAX bead carryover in some wells of my plate. What might be causing this and how can I resolve it?

Some bead carryover is common and has not been documented to affect downstream assays. The most common cause of bead carryover is too much input sample. The best way to determine if the sample amount is the cause of the issue is to run PBS with Xeno (artificial DNA/RNA) in the lysis buffer and then check if there is still bead carryover. If that is not the cause, the initial sample may require special processing, such as bead beating or liquification. If there is still bead carryover without the original sample, and in particular, if the carryover is present in the same wells for multiple runs, then this may be an instrument alignment issue.

Do I need any additional consumables when using MagMAX kits on the MagMAX Express-96 instrument or a KingFisher instrument?

Plastic consumables need to be purchased separately in order to use the MagMAX reagents on the MagMAX Express-96 or a KingFisher instrument. The particular consumables and the amount required will vary by kit and protocol so please check the user guide for the particular MagMAX kit to be sure that all required plastic consumables are available prior to sample processing.

Can I scale up the MagMAX kit reaction volumes? If so, how should I do this?

Yes, reaction volumes for MagMAX kits can be scaled up. In general, the amount of MagMAX reagents should be scaled up proportionally for the increase in sample input amount. Please see the specific kit user guide for any recommendation. If none are included, then optimization may be necessary.

Will addition of BME (2-mercaptoethanol) to the MagMAX lysis buffer cause any issues with nucleic acid extraction?

Addition of BME (2-mercaptoethanol) to the lysis buffer of MagMAX kits is fine. This can be beneficial with more RNase-rich samples like certain types of tissues.
In general, we recommend adding 0.3 µL BME per 100 µL of lysate.