Attune™ Flow Cytometer Syringe
Applied Biosystems™

Attune™ Flow Cytometer Syringe

The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flowRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44520791 mL
447609010 mL
Catalog number 4452079
Price (USD)
579.65
Online Exclusive
610.00
Save 30.35 (5%)
Each
Quantity:
1 mL
Price (USD)
579.65
Online Exclusive
610.00
Save 30.35 (5%)
Each

The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell. Replacement of this syringe is recommended as part of instrument maintenance.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 mL
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeSyringe
Unit SizeEach
Contents & Storage
Store at room temperature