Applied Biosystems™

SOLiD™ EZ Bead™ Emulsifier E20 Reagents Kit

Catalog number:  4452721

 Related applications: PCR | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ EZ Bead™ Emulsifier E20 Reagents Kit contains all necessary reagents and consumables for performing a single E20 EZ Bead Emulsifier run, excluding the EZ Bead™ Oil Pack Kit [PN 4457185]. Please obtain the SOLiD™ EZ Bead™ Oil Pack Kit to have all necessary consumables for an EZ Bead™ System run. One Oil Pack kit is sufficient for 6 x E20 EZ Bead System runs.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer
High Throughput Compatibility: High Throughput-Compatible
Product Line: EZ Bead™, SOLiD™
Product Size: 1 kit
Workflow Step: Emulsion PCR
Yield (up to): 250 million templated beads

Contents & storage

This kit contains 3 boxes:

Box 1: (Store contents at 2C to 8C)
• SOLiD EZ Bead Emulsifier E20 – Aqueous Master Mix
• SOLiD EZ Bead Emulsifier E20 – P1 Beads

Box 2: (Store contents at -15C to -25C)
• SOLiD EZ Bead Emulsifier E20 – P1 Reagent
• SOLiD EZ Bead Emulsifier E20 – P2 Reagent
• SOLiD EZ Bead Emulsifier E20 – BB Solution

Box 3: (Store contents at ambient temperature)
• SOLiD EZ Bead Emulsifier Reagent – 1X TEX Buffer
• SOLiD EZ Bead Emulsifier Reagent – 1X Low TE Buffer

Documents

Manuals & protocols