Applied Biosystems™

SOLiD™ EZ Bead™ Enricher E80 Reagent Kit

Catalog number:  4452725

 Related applications: PCR | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ EZ Bead™ Enricher E80 Reagents Kit contains all necessary reagents for performing a single E80 scale EZ Bead Enricher run.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer
High Throughput Compatibility: High Throughput-Compatible
Product Line: EZ Bead™, SOLiD™
Product Size: 1 kit
Workflow Step: Bead Enrichment
Yield (up to): 1 billion templated beads

Contents & storage

This kit contains 2 boxes:

Box 1: (Store contents at 2C to 8C)
• SOLiD EZ Bead Enricher E80 – Capture Beads
Box 2: (Store contents at -15C to -25C)
• SOLiD EZ Bead Enricher E80 – Capture Oligo

Documents

Manuals & protocols