3-Amino-4-fluorophenylboronic acid, 97%
3-Amino-4-fluorophenylboronic acid, 97%
3-Amino-4-fluorophenylboronic acid, 97%
Thermo Scientific Chemicals

3-Amino-4-fluorophenylboronic acid, 97%

CAS: 873566-75-7 | C6H7BFNO2 | 154.94 g/mol
Have Questions?
Catalog NumberQuantity
4452900101 g
Catalog number 445290010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS873566-75-7
IUPAC Name(3-amino-4-fluorophenyl)boronic acid
Molecular FormulaC6H7BFNO2
InChI KeySYBMNJPUZMUPGQ-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder or solid
Infrared spectrumConforms
HPLC>=96.0 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only