Applied Biosystems™

SOLiD™ EZ Bead™ Enricher Accessories Kit

Catalog number:  4453073

 Related applications: PCR | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This accessories kit contains all customized accessories (tubing, containers, funnels, pouches, filters) for performing E-series scale reactions using the SOLiD™ EZ Bead™ Enricher.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer
Product Line: EZ Bead™, SOLiD™
Product Size: 1 kit
Shipping Condition: Room Temperature
Type: Columns, Filter, Tubes & Vials

Contents & storage

This kit contains the below material: (Store at ambient temperature)

• SOLiD EZ Bead Enricher Column
• SOLiD EZ Bead Enricher Input Tube
• SOLiD EZ Bead Enricher Bead Output Tube
• SOLiD EZ Bead Enricher QC Output Tube
• SOLiD EZ Bead Enricher Hyb Tube
• SOLiD EZ Bead Enricher Capture Oligo Tube
• SOLiD EZ Bead Enricher Release Tube
• SOLiD EZ Bead Enricher Capture Bead Tube
• SOLiD EZ Bead Enricher Unenriched Bead Bottle
• SOLiD EZ Bead Enricher Filter
• SOLiD EZ Bead Enricher Waste Bottle
• SOLiD EZ Bead Sipping Assembly

Documents

Manuals & protocols