Applied Biosystems™
SOLiD™ EZ Bead™ Emulsifier E20 Accessories Kit
Catalog number:  4453076
 Related applications: PCR | SOLiD® Next-Generation Sequencing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
SOLiD™ EZ Bead™ Emulsifier E20 Accessories Kit contains all necessary reagents and consumables for performing a single E20 EZ Bead Emulsifier run, excluding the EZ Bead™ Oil Pack Kit [PN 4457185]. Get the SOLiD™ EZ Bead™ Oil Pack Kit in order to have all necessary consumables for an EZ Bead™ System run. One Oil Pack kit is sufficient for 6 x E20 EZ Bead System runs.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): 5500 Genetic Analyzer
Product Line: EZ Bead™, SOLiD™
Product Size: 1 kit
Type: Containers & Vessels, Tubes & Vials, Tubing
Contents & storage
This kit contains the below material: (Store contents at ambient temperature)
• SOLiD EZ Bead Emulsifier E-Small Jar Pack
• SOLiD EZ Bead Emulsifier E-Small AQ Tube Pack
• SOLiD EZ Bead Emulsifier Tubing Pack
Documents
Manuals & protocols