See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Research and Clinical Analyzers and Instruments
▸
DNA Sequencing Systems
▸
SOLiD™ EZ Bead™ Amplifier E20 Accessories Kit
Applied Biosystems™
SOLiD™ EZ Bead™ Amplifier E20 Accessories Kit
Have Questions?
Catalog Number
Quantity
4453077
4 packs
Catalog number
4453077
Price (USD)
110.00
4 packs
Add to cart
Quantity:
4 packs
Price (USD)
110.00
4 packs
Add to cart
The SOLiD™ EZ Bead™ Amplifier E20 Accessories Kit contains all necessary plastic consumables for performing 4 x E20 EZ Bead Amplifier runs.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
5500 Genetic Analyzer
Product Type
Amplifier E20 Accessories Kit
Quantity
4 packs
Product Line
EZ Bead, SOLiD
Unit Size
4 packs
Contents & Storage
This kit contains one box:
SOLiD™ EZ Bead™ Amplifier Small Accessories Kit⁄box: (Store at ambient)
• SOLiD™ EZ Bead™ Amplifier E-Small Pouch Pack (4 total)
• SOLiD™ EZ Bead™ Amplifier E-Small Funnel Pack (8 total)