Applied Biosystems™

SOLiD™ EZ Bead™ E80 System Consumables

Catalog number:  4453095

 Related applications: PCR | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ EZ Bead™ E80 System Consumables 4-pack contains all necessary reagents and consumables for performing 4 x E80 EZ Bead™ system runs, excluding the EZ Bead™ Oil Pack Kit [PN 4457185]. Get the SOLiD™ EZ Bead™ Oil Pack Kit in order to have all necessary consumables for an EZ Bead™ System run. One Oil Pack kit is sufficient for 2 x E80 EZ Bead System runs.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer
High Throughput Compatibility: High Throughput-Compatible
Product Line: EZ Bead™, SOLiD™
Product Size: 4 pack
Workflow Step: Bead Enrichment, Emulsion PCR
Yield (up to): 1 billion templated beads

Contents & storage

This kit contains the below material: (Storage conditions range from ambient, 2C to 8C and -15C to -25C. Please store according)

• SOLiD EZ Bead Emulsifier E80 Reagent Kit
• SOLiD EZ Bead Emulsifier E80 Accessories Kit
• SOLiD EZ Bead Amplifier E80 Accessories Kit
• SOLiD EZ Bead Enricher E80 Reagent Kit
• SOLiD EZ Bead Enricher Buffer Kit
• SOLiD EZ Bead Enricher E80 Accessories Kit

**NOTE: Sold separately is the SOLiD(tm) EZ Bead(tm) Oil Pack Kit [PN 4457185]

Documents

Manuals & protocols