TaqMan® OpenArray® Real-Time PCR Accessories Kit (automatic shipment enabled)
Catalog number: 4453993
Applied Biosystems™  Related applications: Digital PCR | Genotyping & Genomic Profiling | Real Time PCR (qPCR)
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This kit contains the necessary accessories to process 10 TaqMan OpenArray® Real-Time PCR plates.
TaqMan® OpenArray® Real-Time PCR Plates deliver world-class TaqMan® Assays in a more flexible, high-throughput format. Five customizable plate formats allow you to select from over 61,000 inventoried TaqMan® Gene Expression Assays, which are pre-loaded onto the OpenArray® Plates. View the available formats (PDF). The OpenArray® Real-Time PCR System enables over 8,000 real-time PCR assays to be run simultaneously to generate 32,000+ data points per day.
As part of our greener product alternatives we are discontinuing the shipment of CDs containing TPF files, which are needed to process your TaqMan® OpenArray® Real-Time PCR Plates on the OpenArray® Real-Time PCR System. You will now be able to download your specific TPF and Assay Information Files (AIF) directly.
For Research Use Only. Not for use in diagnostic procedures.
Contents & storage
This kit includes 10 OpenArray cases, 10 immersion fluids, 2 case sealing glues, and 3 frame adhesives. One kit ships automatically with every 10-PK of TaqMan OpenArray Real-Time PCR plates.
Please Note: 5x TaqMan REmix Solution is no longer included in the TaqMan® OpenArray® Real-Time PCR Accessories Kit.
Documents
Manuals & protocols