What are some of the applications for TSP (Transferable Solid Phase)?
Please review the following applications for TSP (Transferable Solid Phase) :
- Reactions such as the ELISA can be performed on the TSP. The pins are coated by submerging in the analyte solution contained in a 96-well plate. Washing and reaction with succeeding antibody or streptavidin conjugates can be performed by transferring the TSP into a washing tray or second 96-well plate filled with the appropriate solution. The TSP is placed into a substrate solution until color is observed and is then removed to ensure a simultaneous start and stop to the enzymatic reaction.
- TSP is available sterile for screening hybridoma cells for the production and secretion of monoclonal antibodies.
- TSP can be used for radioimmunoassays because it can be placed directly on X-ray film and exposed for several hours. Only the tips of the pins should be incubated with the radiolabeled reagent.
- TSP can be used with the OmniTray for performing dot blots and for replicating bacterial clones from a 96-well plate.
- Reactions such as the ELISA can be performed on the TSP. The pins are coated by submerging in the analyte solution contained in a 96-well plate. Washing and reaction with succeeding antibody or streptavidin conjugates can be performed by transferring the TSP into a washing tray or second 96-well plate filled with the appropriate solution. The TSP is placed into a substrate solution until color is observed and is then removed to ensure a simultaneous start and stop to the enzymatic reaction.
- TSP is available sterile for screening hybridoma cells for the production and secretion of monoclonal antibodies.
- TSP can be used for radioimmunoassays because it can be placed directly on X-ray film and exposed for several hours. Only the tips of the pins should be incubated with the radiolabeled reagent.
- TSP can be used with the OmniTray for performing dot blots and for replicating bacterial clones from a 96-well plate.