Product Image
Applied Biosystems™

TaqMan™ RNase P Instrument Verification Plate for ViiA™ 7 System, 384-well

Used to perform verification of your ViiA™7 System with 384 Well Block.
Have Questions?
Catalog NumberQuantity
44552801 x 384-well plate
Catalog number 4455280
Price (EUR)
991,10
Special offer
Online exclusive
Ends: 27-Mar-2026
1.802,00
Save 810,90 (45%)
Each
Quantity:
1 x 384-well plate
Price (EUR)
991,10
Special offer
Online exclusive
Ends: 27-Mar-2026
1.802,00
Save 810,90 (45%)
Each

Used to perform verification of your ViiA™7 System with 384 Well Block.

Please see your instrument maintenance guide for full instructions on use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeFAM
Calibration TypeRNase P Calibration, Instrument Verification Plate
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA™ 7 System, QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio™ 12k Flex
Format384-well Plate
Product LineTaqMan, ViiA
Quantity1 x 384-well plate
Shipping ConditionDry Ice
Product TypeRnase P Instrument Verification Plate
Unit SizeEach
Contents & Storage
Store Calibration Plates at -15°C to -25°C

Frequently asked questions (FAQs)

When do I need to run the RNaseP calibration?

The RNaseP verification plate contains template, master mix, and a TaqMan assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

When do I need to run the RNaseP calibration for the QuantStudio 12K Flex Real-Time PCR System?

The RNaseP verification plate contains template, master mix, and a TaqMan assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

I cannot open the QuantStudio 3/5 Design & Analysis 2 (DA2) software on my Mac. What are the software requirements for Mac?

To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:

Safely open apps on your mac

Please note, the software should run on Mac OS 10.10 or later.

Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Real-Time PCR Software?

If you are analyzing results from the ViiA 7 Real-Time PCR System, QuantStudio 6 Flex Real-Time PCR System, or QuantStudio 7 Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Real-Time PCR Software following the instructions below:
1. Open the data file in the QuantStudio Real-Time PCR Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.