2-(Tetrazol-5-yl)phenylboronic acid, 95%
2-(Tetrazol-5-yl)phenylboronic acid, 95%
2-(Tetrazol-5-yl)phenylboronic acid, 95%
2-(Tetrazol-5-yl)phenylboronic acid, 95%
Thermo Scientific Chemicals

2-(Tetrazol-5-yl)phenylboronic acid, 95%

CAS: 155884-01-8 | C7H7BN4O2 | 189.97 g/mol
Have Questions?
Catalog NumberQuantity
4455500101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445550010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS155884-01-8
IUPAC Name[2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
Molecular FormulaC7H7BN4O2
InChI KeyGVRXWYFECKHTSJ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1C1=NNN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94.0 %
Infrared spectrumConforms
Appearance (Form)Solid
Additional infoMay contain varying amounts of anhydrides
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only