Nitromethane, 96%, for HPLC
Nitromethane, 96%, for HPLC
Nitromethane, 96%, for HPLC
Thermo Scientific Chemicals

Nitromethane, 96%, for HPLC

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445670025
Price (USD)
283.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
283.00
Each
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Water=<0.03 %
UVat 386 nm A: =<0.5
UVat 405 nm A: =<0.05
GC>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only