Applied Biosystems™
SOLiD™ EZ Bead™ Emulsifier Oil Pack Kit
Catalog number:  4457185
 Related applications: PCR | SOLiD® Next-Generation Sequencing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The SOLiD™ EZ Bead™ Emulsifier Oil Pack kit contains the necessary reagents and plastics accessories to create the oil phase. The kit accommodates E10, E20, and E80 reaction scales.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): 5500 Genetic Analyzer
High Throughput Compatibility: High Throughput-Compatible
Product Line: EZ Bead™, SOLiD™
Product Size: 1 kit
Workflow Step: Emulsion PCR
Contents & storage
This kit contains the below material:
SOLiD EZ Bead Emulsifier Oil Kit: (Store contents at ambient temperature)
• SOLiD EZ Bead Emulsifier Oil Kit – Stabilizer 1
• SOLiD EZ Bead Emulsifier Oil Kit – Stabilizer 2
• SOLiD EZ Bead Emulsifier Oil Kit – Oil
SOLiD EZ Bead Emulsifier Oil Accessory: (Store contents at ambient temperature)
• SOLiD EZ Bead Emulsifier Oil Kit– Oil Mix Bottle
• SOLiD EZ Bead Emulsifier Oil Kit– Oil Funnel
Documents
Manuals & protocols