Applied Biosystems™

OpenArray™ AccuFill™ System

Catalog number:  4457243

Related applications:

Real Time PCR (qPCR)

Back to top

Description

This is the AccuFill™ System for the OpenArray® Real-Time PCR Platform.

The OpenArray® Real-Time PCR Instrument and OpenArray® AccuFill™ System combination can process hundreds to thousands of samples per day, enabling mid-density, high-throughput results with ease of use.

OpenArray® Real-Time PCR Components
The platform includes the OpenArray® Real-Time PCR Instrument, computer⁄monitor, analysis software for genotyping and real-time applications, installation, training and OpenArray® Real-Time PCR Platform warranty.

The OpenArray® Real-Time PCR Platform includes:
• OpenArray® AccuFill™ System—Accurately loads samples onto OpenArray® Plates
• OpenArray® Case Sealing Station—Seals OpenArray® Plates in cases
• OpenArray® Real-Time PCR Software and OpenArray® Genotyping Software—Acquire, analyze, and manage data quickly and easily

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: OpenArray™ Plate
Product Line: AccuFill™, OpenArray™
Includes: Real Time PCR Instrument
For Use With (Equipment): OpenArray™
Product Type: Open Array Accu Fill System

Contents & storage

The AccuFill™ System can be kept at ambient temperature.