Product Image
Invitrogen™

Ambion™ In Vivo Negative Control #1 siRNA

Ambion™ In Vivo siRNA Controls• Validated siRNA controls for optimizing siRNA experiments targeting coding and noncoding transcripts• Positive Control siRNAs자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
44572875 nmol
445940550 nmol
4457289250 nmol
카탈로그 번호 4457287
제품 가격(KRW)
572,000
온라인 행사
Ends: 30-Sep-2026
653,000
할인액 81,000 (12%)
Each
수량:
5 nmol
제품 가격(KRW)
572,000
온라인 행사
Ends: 30-Sep-2026
653,000
할인액 81,000 (12%)
Each
Ambion™ In Vivo siRNA Controls
• Validated siRNA controls for optimizing siRNA experiments targeting coding and noncoding transcripts
• Positive Control siRNAs functionally tested in several common cell lines
• Negative Controls functionally proven to have minimal effects on cell proliferation and viability
• Include Silencer™ Select modifications for enhanced specificity or Ambion™ In Vivo modifications for increased stability
• For use in human, mouse, and rat cells

What are Ambion™ In Vivo siRNAs?
Ambion™ In Vivo siRNAs are designed using the proven Silencer™ Select algorithm and incorporate chemical modifications for superior serum stability with in vivo delivery. The added serum stability does not compromise the performance of the siRNAs. Ambion™ In Vivo siRNAs have been shown to be non-toxic in vivo (mouse) and non-immunogenic (cell-based assays). In cell-based assays, Ambion™ In Vivo siRNAs exhibit equivalent or better potency than Silencer™ Select siRNAs. The result is better confidence in your in vivo RNAi experiments, better knockdown when paired with an efficient delivery solution, and the assurance of limited non-specific effects.

Negative Control siRNAs
Negative control siRNAs—siRNAs with sequences that do not target any gene product—are essential for determining the effects of siRNA delivery and for providing a baseline to compare siRNA-treated samples. Ambion™ In Vivo siRNAs and have no significant sequence similarity to mouse, rat, or human gene sequences. These negative control siRNAs have been tested by microarray analysis and shown to have minimal effects on gene expression. In addition, these negative control siRNA has been tested in multi-parametric cell-based assays and are proven to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested.

Quality Control
Ambion™ synthesizes and purifies each Ambion™ In Vivo siRNA in state-of-the-art facilities to meet the highest quality standards. As part of our rigorous quality control procedures, each RNA oligonucleotide is analyzed by MALDI-TOF mass spectrometry, and analytical HPLC is used to monitor purity. To provide the utmost in quality, we also assess each annealed siRNA by gel electrophoresis to confirm that the strands anneal properly. The result is premium-quality siRNA that is purified and ready to use.

In Vivo Ready Controls
Our “In Vivo Ready” Control siRNAs are subjected to that extra level of purification and testing required for the introduction of siRNAs into animals. After HPLC purification and annealing, each siRNA is further purified utilizing a process that removes excess salt via a semi-permeable membrane. The result is highly pure siRNA with minimal salt content, suitable for in vivo applications. In vivo siRNAs are then sterile filtered, and tested for the presence of endotoxin. At concentrations of 50 μM in presence of deionized water, in vivo Ready siRNAs contain less than 5.0 mM Na+, less than 0.06 mM K+, and less than 0.02 mM Mg2+.
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)RNAi, Transfection
라벨 또는 염료Unconjugated
제품라인Ambion
제품 유형siRNA
순도In Vivo, HPLC
수량5 nmol
배송 조건Room Temperature
제어 유형Negative Control
RNAi TypesiRNA
Unit SizeEach
구성 및 보관
The siRNA (annealed) is shipped at ambient temperature as dried down oligonucleotides in a single tube, along with 1.75 ml Nuclease-free Water for resuspension. Store at or below –20°C.

자주 묻는 질문(FAQ)

What are the benefits of using our Invitrogen In vivo siRNA?

- 100x more stable than unmodified siRNA - longer knockdown in vivo
- Compatible with any 21-mer sequence - no loss of potency
- Non-toxic - actually reduces siRNA sequence specific immune responses
- Enhanced specificity - up to 90% reduction in off target effects
- Both Pre-designed and Validated Invitrogen In Vivo siRNAs offered


Find additional tips, troubleshooting help, and resources within our RNAi Support Center.

What siRNA do you offer that I can use for my in vivo studies, and what are the differences between them?

Please see the different siRNA we offer for in vivo studies:

Ambion In Vivo Pre-designed siRNAs are designed using the Silencer Select algorithm and incorporate additional chemical modifications for superior serum stability (half life >5 hours at 37 degrees C in 90% mouse serum) with in vivo applications. Ambion In Vivo siRNAs are non-toxic and non-immunogenic in vitro (peripheral blood nononuclear cells; PBMC) and in vivo (mouse). In cell-based assays, Ambion In Vivo siRNAs exhibit potency and specificity equivalent to Silencer Select siRNAs.
Ambion In Vivo Custom Designed siRNAs are designed to your specified target, using the Silencer Select algorithm and Ambion In Vivo chemical modifications.
Ambion In Vivo Custom Synthesized siRNAs are synthesized with your sequence, with Ambion In Vivo chemical modifications for serum stability.

Our Ambion In Vivo siRNA can be ordered with the following purities:

Desalted: Desalted and analyzed by MALDI-TOF mass spectrometry.
HPLC: HPLC purified and analyzed by MALDI-TOF and analytical HPLC
In Vivo Ready : HPLC purified, analyzed by MALDI-TOF and analytical HPLC, dialyzed to remove salts, sterile filtered, and endotoxin tested

Use our GeneAssist tool to order your custom siRNA (http://www.thermofisher.com/order/custom-genomic-products/tools/sirna/).

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.