Applied Biosystems™

SOLiD™ Buffer D, Double

Catalog number:  4458935

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ Buffer D, Double is used to reset primers for each tag sequenced on the 5500 or 5500xl Genetic Analyzers. The buffer contains sufficient material to reset 216 primers, or 18 primers per lane.

The SOLiD™ Buffers are used for ligation sequencing on the 5500⁄5500xl Genetic Analyzer. The “Main" kits are the primary kits necessary for typical sequencing runs.

Double-sized buffers have sufficient reagents for covering two FlowChips.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: SOLiD™
Product Size: 1 kit
Recommended Applications: De Novo Sequencing, Shotgun Sequencing
Shipping Condition: Wet Ice

Contents & storage

There is sufficient buffer for two FlowChips.

Store buffer at 2°C to 8°C.

Documents

Manuals & protocols