Applied Biosystems™

SOLiD™ FWD Buffer, Double

Catalog number:  4459193

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ FWD Buffer, Double is used for fragment, mate-paired, and paired-end runs, for ligation sequencing in the forward direction. This buffer must be used with the SOLiD™ FWD SR Kit. This buffer contains sufficient material to run 900 to 1212 cycles, or up to 101 cycles per lane.

The SOLiD™ Buffers are used for ligation sequencing on the 5500⁄5500xl Genetic Analyzer. The “Main" kits are the primary kits necessary for typical sequencing runs.

Double-sized buffers have sufficient reagents for covering two FlowChips.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: SOLiD™
Product Size: 1 kit
Recommended Applications: De Novo Sequencing, Shotgun Sequencing
Shipping Condition: Wet Ice

Contents & storage

There is sufficient buffer for two FlowChips.

Store buffer at 2°C to 8°C .

Documents

Manuals & protocols