MycoSEQ™ Mycoplasma Detection Kit, with PrepSeq™ sample preparation, 100 Reactions - FAQs
MycoSEQ™ Mycoplasma Detection Kit, with PrepSeq™ sample preparation, 100 Reactions - FAQs
View additional product information for MycoSEQ™ Mycoplasma Detection Kits - FAQs (4460623, 4460626)
1 product FAQs found
How can I remove mycoplasma contamination from my cell culture medium?
Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.