Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%

CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 446081000
Price (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10378-23-1
IUPAC Name2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
Molecular FormulaC10H20N2Na4O10
InChI KeyDGWOWDJDIBPYOR-UHFFFAOYSA-N
SMILESO.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying6.4 % to 10.6 %
Titration Complexometric99.0 to 102.0 % (on dry substance)
Appearance (Form)Crystalline powder
Solubility(16% in water) Clear solution
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only