TaqMan™ Multiplex Master Mix
TaqMan™ Multiplex Master Mix
TaqMan™ Multiplex Master Mix
Applied Biosystems™

TaqMan™ Multiplex Master Mix

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorterRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44618811 mL
44618825 mL
44618842 x 5 mL
44842625 x 5 mL
448426310 x 5 mL
448629550 mL
Catalog number 4461881
Price (USD)
224.65
Online exclusive
244.00
Save 19.35
Each
Quantity:
1 mL
Price (USD)
224.65
Online exclusive
244.00
Save 19.35
Each
Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (<40 minutes) with excellent concordance of results in singleplex and multiplex reactions. TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA PolymeraseUracil-N Glycosylase (UNG), dNTPs with dUTP, MUSTANG PURPLE™ dye (passive reference), and optimized buffer components. It is supplied at a 2X concentration.

Features of TaqMan™ Multiplex Master Mix include:

• Optimized for multiplexing—validated for quadplex gene expression and genotyping assays
• Best-in-class performance—superior sensitivity, accuracy, dynamic range, and specificity
• Reduced run times on fast and standard instrumentation—optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
• Seamless integration into your experiments—validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
• Carry-over contamination control—contains heat-labile UDG
• Broad instrument compatibility

Optimized for Multiplexing
Confidence is paramount when it comes to your results. TaqMan™ Multiplex Master Mix is designed to deliver accurate results for multplex reactions and ensure excellent concordance of results in both singleplex and multiplex reactions.

Best-in-class Performance
TaqMan™ Multiplex Master Mix is designed for performance equal to or better than other multiplex master mixes on the market. TaqMan™ Multiplex Master Mix has been benchmarked against the leading suppliers' multiplex master mixes to help ensure that it succeeds in providing best-in-class sensitivity, accuracy, dynamic range, and specificity.

Reduced Run Times on Standard Instrumentation
TaqMan™ Multiplex Master Mix has been optimized for use with both fast and standard instrumentation, enabling researchers who currently own standard instruments to also reap the performance benefits and time savings this mix provides.

Carry-over Contamination Control
TaqMan™ Multiplex Master Mix contains heat-labile uracil-DNA glycosylase (UDG) for worry-free carryover contamination control. The dUTP in the mix ensures that any amplified DNA will contain uracil, while UDG removes uracil residues from single- or double-stranded DNA, preventing dU-containing DNA from serving as template in future PCRs. A UDG incubation step before PCR cycling destroys any contaminating dU-containing product from previous reactions. UDG is then inactivated by the high temperatures during normal PCR cycling, thereby allowing the amplification of genuine target sequences.

Instrument Compatibility
TaqMan™ Multiplex Master Mix can be used in fast cycling mode on both standard and fast qPCR instruments, such as: 7500 Real-Time PCR System, 7500 Fast Real-Time PCR System, ViiA™ 7 Real-Time PCR System, and QuantStudio™ 6 Flex, 7 Flex, and 12K Flex Real-Time PCR systems.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
No. of Reactions100 Reactions
PolymeraseAmpliTaq Fast DNA Polymerase
Product LineTaqMan
Product TypeMultiplex Master Mix
Quantity1 mL
Sample TypeDNA
Sufficient For100 Reactions
Volume1 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Virus Detection
GC-Rich PCR PerformanceHigh
PCR MethodMultiplex qPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
Contains 1 tube (1 mL) of 2X TaqMan™ Multiplex Master Mix sufficient for 100 reactions.


TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer.

Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

I need to multiplex but cannot use ROX. What options do I have?

For multiplex assays that include reporter dyes overlapping the ROX channel (i.e. JUN), we recommend using one of our optimized multiplex master mixes that include the passive reference dye, MUSTANG PURPLE. MUSTANG PURPLE is compatible with QuantStudio 5, 6, 7, and 12K Flex, 7500, 7500 Fast, and ViiA 7 real-time PCR instruments.

While not ideal, we also offer master mixes that contain no passive reference dyes. For selection help for master mixes with no passive reference, please contact Technical Support at techsupport@thermofisher.com.

What master mix should I use for multiplexing in my qPCR experiments?

The choice of master mix depends upon the number of assays in your reaction, the application, and instrument compatibility. Please refer to our real-time PCR master mixes and instrument compatibility chart (https://assets.thermofisher.com/TFS-Assets/LSG/brochures/master-mix-instrument-compatibility.pdf) prior to selecting one of the master mixes below:

Gene Expression Assays:
- Single, duplex, and triplex assays: TaqMan Fast Advanced Master Mix or TaqMan Fast Virus 1-Step Master Mix
- Multiplexing up to 4 assays: TaqMan Multiplex Master Mix or TaqPath 1-Step Multiplex Master Mix

SNP Genotyping Assays:
- Single SNP Genotyping assay: TaqPath ProAmp Master Mix
- Duplex SNP Genotyping assays: TaqPath ProAmp Multiplex Master Mix, TaqMan Multiplex Master Mix

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.