SOLiD™ FlowChip Deposition Buffer 1

Catalog number: 4462407

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SOLiD™ FlowChip Deposition Buffer 1 is required for depositing templated beads onto a SOLiD™ FlowChip for sequencing on the 5500⁄5500xl Genetic Analyzer.

Store the SOLiD™ FlowChip Deposition Buffer 1 at 2°C to 8°C until ready for use.

The SOLiD™ FlowChip Deposition Buffer 1 is also included in the SOLiD™ FlowChip Deposition Kit.

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: SOLiD™
Product Size: 1 bottle
Shipping Condition: Wet Ice

Contents & storage

1 bottle, Store at 2° C to 8° C

Documents

Manuals & protocols