3,3-Dimethoxypropanenitrile, 90%
3,3-Dimethoxypropanenitrile, 90%
3,3-Dimethoxypropanenitrile, 90%
Acros Organics

3,3-Dimethoxypropanenitrile, 90%

CAS: 57597-62-3 | C5H9NO2 | 115.13 g/mol
Have Questions?
Catalog NumberQuantity
446260250
also known as 44626
25 mL
Catalog number 446260250
also known as 44626
HSN 29269000
Price (INR)
34,427.00
Each
Quantity:
25 mL
Request bulk or custom format
Price (INR)
34,427.00
Each
Chemical Identifiers
CAS57597-62-3
IUPAC Name3,3-dimethoxypropanenitrile
Molecular FormulaC5H9NO2
InChI KeyJYHSJQNYYLGMEI-UHFFFAOYSA-N
SMILESCOC(CC#N)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4120 to 1.4160 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Colorless to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only