1-Propanol, specified according to the requirements of Ph.Eur.
1-Propanol, specified according to the requirements of Ph.Eur.
1-Propanol, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

1-Propanol, specified according to the requirements of Ph.Eur.

CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 446270025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Boiling point96°C to 98°C (760 mmHg)
UVat 230 nm A: =<0.300
Non-volatile matter=<0.004 %
Refractive index1.3840 to 1.3870 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only