Ion Torrent™

Partek™ Genomics Suite™ Software Academic License, 12 Months

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Partek® Genomics Suite™ Software is a full-featured software package for the analysis, comparison, and integration of genomic data. This product, which can be ordered through our website, is supplied directly by Partek Incorporated.

Partek® Genomics Suite™ Software gives you:

• Sophisticated capabilities for DNA-seq, ChIP-seq, and RNA-seq experiments
• Statistical analysis of each base pair for DNA-seq
• Extensive annotations
• SFF, FASTQ, and SAM/BAM file formats

Partek® Genomics Suite™ Software—Your Start-To-Finish Solution For Integrated Genomics
Partek® Genomics Suite™ Software offers a variety of next generation sequencing workflows including DNA-seq, ChIP-seq and RNA-seq. Within the DNA-seq workflow, Genomics Suite™ Software extends the analysis, identifying DNA variations beyond simple filters based on depth of coverage or allele frequencies. Genomics Suite™ Software performs a statistical test at each base pair to determine if the variation seen at that base is statistically different from the control, while users have the ability to change the definition of "control." Comparisons can be made in a paired manner; for example, a single sample can be compared against a control, variants can be identified against a consensus, or variants can be found across a collection of samples without a defined reference. Genomics Suite™ Software offers integration using the industry standard file formats SFF, FASTQ, and SAM/BAM. Once variants have been identified, a collection of rich annotations allows the user to explore the biology that lies underneath the variation.

Now Integrated with TaqMan® Assays
Partek® Genomics Suite™ Software is now integrated with TaqMan® real time PCR assays. The software allows you to easily select the real time PCR assays to validate your next generation sequencing data.

This Partek® Genomics Suite™ product is manufactured and offered by Partek Incorporated. Purchases of this product will be processed by us on behalf of Partek, as will all subsequent fulfillment, shipment, and support needs.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion PGM™ System, Ion Proton™ System
Software Type: Sequencing Data Analysis
Data Input: BAM, FASTQ, SAM, SFF
License Type: Academic
Operating System: CentOS, Fedora, Red Hat, Ubuntu, Windows 7, Windows 8, Windows XP SP3
Product Line: Partek®
Product Size: 1 each
Software Category: Software
Shipping Condition: Room Temperature

Documents

Manuals & protocols